Meclofenamic acid sodium salt ≥99%

Supplier: Thermo Scientific Chemicals

J60484-03 J60484-06
CAAAJ60484-03EA 79.47 CAD
CAAAJ60484-03 CAAAJ60484-06
Meclofenamic acid sodium salt ≥99%
Meclofenamic acid sodium salt

Inhibits 5-lipoxygenase and cyclooxygenase; NSAID


Crystalline solid


Applications: Inhibits 5-lipoxygenase and cyclooxygenase; NSAID

Formula: C₁₄H₁₀Cl₂NNaO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00077376
CAS Number: 6385-02-0
EINECS: 228-983-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR