Calpeptin ≥98%

Supplier: Thermo Scientific Chemicals

J60481-MA J60481-MF
CAAAJ60481-MAEA 722.27 CAD
CAAAJ60481-MA CAAAJ60481-MF
Calpeptin ≥98%
Calpeptin

Inhibitor of the calcium-dependent protease calpain and cathepsin L


Powder


Applications: Inhibitor of the calcium-dependent protease calpain and cathepsin L

Formula: C₂₀H₃₀N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00155623
CAS Number: 117591-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR