Hexamethonium bromide ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N1,N1,N1,N6,N6,N6-hexamethylhexane-1,6-diaminium bromide, Hexane-1,6-bis(trimethylammonium Bromide), Hexamethylenebis(trimethylammonium Bromide)
J60466-14
J60466-22
CAAAJ60466-14EA
163.23
CAD
CAAAJ60466-14
CAAAJ60466-22
Hexamethonium bromide ≥98%
Hexamethonium bromide
Nicotinic acetyl choline receptor antagonist
Powder
Applications: Nicotinic acetyl choline receptor antagonist
Formula:
C₁₂H₃₀Br₂N₂ MW: 362.19 g/mol Melting Pt: 282…285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011787 CAS Number: 55-97-0 EINECS: 200-249-7 Merck Index: 14,4688 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...