Phosphate-buffered saline (PBS, 10X), no potassium, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60465-AP
J60465-K2
CAAAJ60465-APEA
150.07
CAD
CAAAJ60465-AP
CAAAJ60465-K2
Phosphate-buffered saline (PBS, 10X), no potassium, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00131855
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...