Tioconazole ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-((2-Chlorothiophen-3-yl)methoxy)-2-(2,4-dichlorophenyl)ethyl)-1H-imidazole

J60459-03 J60459-06
CAAAJ60459-03EA 223.27 CAD
CAAAJ60459-03 CAAAJ60459-06 CAAAJ60459-14
Tioconazole ≥98%
Tioconazole

Potent inhibitor of cytochrome-P450


Powder


Applications: Potent inhibitor of cytochrome-P450

Formula: C₁₆H₁₃Cl₃N₂OS
MW: 387.71 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00057276
CAS Number: 65899-73-2
EINECS: 265-973-8
Merck Index: 14,9452

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR