Chlorogenic acid

Supplier: Thermo Scientific Chemicals

J60457-03 J60457-06 J60457-MD
CAAAJ60457-03EA 215.7 CAD
CAAAJ60457-03 CAAAJ60457-06 CAAAJ60457-MD
Chlorogenic acid
Chlorogenic acid

An analog of caffeic acid


Powder


Applications: An analog of caffeic acid

Formula: C₁₆H₁₈O₉
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003862
CAS Number: 327-97-9
EINECS: 206-325-6
Merck Index: 14,2142

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR