Myricetin 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

J60450-MA J60450-MB J60450-MF
CAAAJ60450-MAEA 143.97 CAD
CAAAJ60450-MA CAAAJ60450-MB CAAAJ60450-MF
Myricetin 98%
Myricetin

Common flavanol in plants that inhibits phenolsulfotransferase. Strongly inhibits yeast alpha-glucosidase


Crystalline solid


Applications: Common flavanol in plants that inhibits phenolsulfotransferase. Strongly inhibits yeast alpha-glucosidase

Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD00006827
CAS Number: 529-44-2
EINECS: 208-463-2
Merck Index: 14,6332

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR