TRIS-buffered saline (TBS, 10X), with 0.5% Tween 20, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60448-AP
J60448-K2
J60448-K3
CAAAJ60448-APEA
121.41
CAD
CAAAJ60448-AP
CAAAJ60448-K2
CAAAJ60448-K3
TRIS-buffered saline (TBS, 10X), with 0.5% Tween 20, Thermo Scientific
Buffers
Electrophoresis Buffers
Washing Buffers
Viscous liquid
Physical Form: Viscous liquid
Appearance: Colorless
MDL No.: MFCD00678198
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...