Sodium chloride 5 M in aqueous solution RNase-free

Supplier: Thermo Scientific Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

J60434-AK J60434-AE
CAAAJ60434-AKEA 150.07 CAD
CAAAJ60434-AK AAJ60434-AE
Sodium chloride 5 M in aqueous solution RNase-free
Sodium chloride

Liquid


CAS No.: 7647-14-5 Molecular Formula: ClNa Formula Weight: 58.44 Physical Form: Liquid MDL No.: MFCD00003477

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR