Carboplatin

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)

J60433-03 J60433-06
CAAAJ60433-03EA 1145.75 CAD
CAAAJ60433-03 CAAAJ60433-06
Carboplatin
Carboplatin

An antitumor agent


Powder


Applications: An antitumor agent

Formula: C₆H₁₂N₂O₄Pt
MW: 371.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00070464
CAS Number: 41575-94-4
EINECS: 255-446-0
Merck Index: 14,1822

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR