Quinidine sulfate Dihydrate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol sulphate hydrate(2:1:2), (S)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol sulfate hydrate(2:1:2)

J60426-06 J60426-14
CAAAJ60426-06EA 167.32 CAD
CAAAJ60426-06 CAAAJ60426-14
Quinidine sulfate Dihydrate ≥98%
Quinidine sulfate Dihydrate

A sodium channel blocker, and to a lesser extent also blocks potassium channels


Soluble in chloroform, water (11 mg/ml at 20°C), alcohol, and ethanol (50 mg/ml).


Applications: A sodium channel blocker, and to a lesser extent also blocks potassium channels

Formula: C₄₀H₄₈N₄O₄.H₂O₄S·2H₂O
MW: 782.94 g/mol
Melting Pt: 212…214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149346
CAS Number: 6591-63-5
EINECS: 200-046-3
Merck Index: 14,8060

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR