Doxycycline hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J60422-06 J60422-14 J60422-03
CAAAJ60422-06EA 225.36 CAD
CAAAJ60422-06 CAAAJ60422-14 AAJ60422-03
Doxycycline hydrochloride
Doxycycline hydrochloride

Inhibits angiogenesis and reduces lung metastases by inhibiting matrix metalloproteinases


Crystalline powder


Applications: Inhibits angiogenesis and reduces lung metastases by inhibiting matrix metalloproteinases

Formula: C₂₂H₂₄N₂O₁₀·HCl
Melting Pt: 195…201 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427564
CAS Number: 10592-13-9
EINECS: 234-198-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR