Epirubicin hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (8S,10S)-10-(((2R,4S,5R,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride, Epirubicin HCl

J60411-MA J60411-MC
CAAAJ60411-MAEA 470.98 CAD
CAAAJ60411-MA CAAAJ60411-MC
Epirubicin hydrochloride
Epirubicin hydrochloride

Cell-permeable anthracycline anti-tumor antibiotic


Solid


Applications: Cell-permeable anthracycline anti-tumor antibiotic

Formula: C₂₇H₂₉NO₁₁·HCl
MW: 579.98 g/mol
Melting Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00941448
CAS Number: 56390-09-1
EINECS: 260-145-2
Merck Index: 14,3623

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR