Sodium bicarbonate, 1 M buffer soln., pH 8.5, Thermo Scientific

Supplier: Thermo Scientific
J60408-AP J60408-AP
CAAAJ60408-APEA 225.1 CAD
AAJ60408-AP CAAAJ60408-AP
Sodium bicarbonate, 1 M buffer soln., pH 8.5, Thermo Scientific
Buffers Biological Buffers
Liquid

CAS No.: 144-55-8
Physical Form: Liquid
MDL No.: MFCD00003528
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR