Formaldehyde 4% in PBS stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
Stabilization:
Stabilised with methanol
J60401-AK
J60401-AP
CAAAJ60401-AKEA
105.23
CAD
CAAAJ60401-AK
CAAAJ60401-AP
Formaldehyde 4% in PBS stabilised
Formaldehyde
Formaldehyde 4% (= 10% Formaline solution)
Storage Temperature: 2°C to 8°C
Appearance: Colourless
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...