Potassium oxonate

Supplier: Thermo Scientific Chemicals

Synonyms: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

J60400-06 J60400-14 J60400-22
CAAAJ60400-06EA 88.57 CAD
CAAAJ60400-06 CAAAJ60400-14 CAAAJ60400-22
Potassium oxonate
Potassium oxonate

Inhibitor of uricase


Powder


Applications: Inhibitor of uricase

Formula: C₄H₃N₃O₄
MW: 195.18 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00010565
CAS Number: 2207-75-2
EINECS: 218-627-5
Merck Index: 14,6947

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR