Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

J60384-LB0 J60384-MB
CAAAJ60384-LB0EA 250.98 CAD
CAAAJ60384-LB0 CAAAJ60384-MB
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) 98%
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Aminopeptidase B inhibitor. Also inhibits leucine aminopeptidase.


Powder


Aminopeptidase B inhibitor. Also inhibits leucine aminopeptidase

Formula: C₁₆H₂₅ClN₂O₄
MW: 308.4 · 36.5 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00058004
CAS Number: 65391-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR