Phosphate, 0.2 M buffer soln., pH 7.4, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60380-AP
J60380-K2
CAAAJ60380-APEA
150.07
CAD
CAAAJ60380-AP
CAAAJ60380-K2
Phosphate, 0.2 M buffer soln., pH 7.4, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...