Bortezomib 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ((R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butyl)boronic acid

J60378-MA J60378-MB
CAAAJ60378-MAEA 597.71 CAD
CAAAJ60378-MA CAAAJ60378-MB
Bortezomib 99%
Bortezomib

A potent, highly selective and reversible inhibitor of the 20S proteasome


A potent, highly selective and reversible inhibitor of the 20S proteasome


A potent, highly selective and reversible inhibitor of the 20S proteasome

Formula: C₁₉H₂₅BN₄O₄
Melting Pt: 139…143 °C
Storage Temperature: Freezer
MDL Number: MFCD09056737
CAS Number: 179324-69-7
UN: 3249
ADR: "6.1",III
Merck Index: 14,1351

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR