Ammonium sulfate 0.5 M in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

J60368-AD J60368-AE
CAAAJ60368-ADEA 50.4 CAD
CAAAJ60368-AD CAAAJ60368-AE
Ammonium sulfate 0.5 M in aqueous solution
Ammonium sulphate

Liquid


CAS No.: 7783-20-2 Molecular Formula: H8N2O4S Formula Weight: 132.14 Physical Form: Liquid MDL No.: MFCD00003391

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR