o-Phenylenediamine dihydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzene-1,2-diamine dihydrochloride, 1,2-Phenylenediammonium dichloride, 1,2-Diaminobenzene dihydrochloride, 1,2-Phenylenediamine dihydrochloride

J60354-22 J60354-14
CAAAJ60354-22EA 437.66 CAD
CAAAJ60354-22 CAAAJ60354-14
o-Phenylenediamine dihydrochloride 98+%
o-Phenylenediamine dihydrochloride

A peroxidase substrate for ELISA procedures


Powder


A peroxidase substrate for ELISA procedures

Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 205 °C
Storage Temperature: Freezer
MDL Number: MFCD00012966
CAS Number: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR