Sodium cacodylate, 0.1 M buffer soln., pH 7.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60344-AD
J60344-AE
CAAAJ60344-EQEEA
139.4
CAD
CAAAJ60344-EQE
AAJ60344-AD
AAJ60344-AE
Sodium cacodylate, 0.1 M buffer soln., pH 7.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 6131-99-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00149079
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...