Clofibrate 95

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate

J60342-03 J60342-06 J60342-14 J60342-22
CAAAJ60342-03EA 41.71 CAD
CAAAJ60342-03 CAAAJ60342-06 CAAAJ60342-14 CAAAJ60342-22
Clofibrate 95
Clofibrate

Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450’s CYP4A1 and CYP4A3.


Purity: 95+% CAS No.: 637-07-0 Molecular Formula: C12H15ClO3 Formula Weight: 242.70 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.14 Refractive Index: 1.5 Appearance: Clear Boiling Point: 154-156° Beilstein Reference: 1913459 UN No.: 3082 MDL No.: MFCD00000615


A lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL.

Formula: C12H15ClO3
MW: 242.70 g/mol
Boiling Pt: 154…156 °C
Density: 1.14
Flash Pt: 113 °C (235 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000615
CAS Number: 637-07-0
EINECS: 211-277-4
UN: 3082
ADR: 9 III
Merck Index: 14,2377

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR