MOPS, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60328-AE J60328-AK J61236-AK J62368-AK J62839-AE J62839-AK J63369-AE J63369-AK
CAAAJ60328-AEEA 116.51 CAD
CAAAJ60328-AE CAAAJ60328-AK CAAAJ60922-AE CAAAJ60922-AK CAAAJ61236-AE CAAAJ61236-AK CAAAJ62368-AE CAAAJ62368-AK CAAAJ62476-AE CAAAJ62476-AK CAAAJ62477-AE CAAAJ62477-AK CAAAJ62839-AE CAAAJ62839-AK CAAAJ63369-AE CAAAJ63369-AK
MOPS, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 1132-61-2
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00006183

Order Now

SPECIFICATIONS

Environmentally Preferable
Odour/scent Odorless Odorless
Physical form
CAS No.
MDL No. MFCD00006183
Storage temperature 2°C to 8°C 2°C to 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR