TAPS, 0.2 M buffer soln., pH 8.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60322-AE J60322-AK
CAAAJ60322-AEEA 170.01 CAD
CAAAJ60322-AE CAAAJ60322-AK
TAPS, 0.2 M buffer soln., pH 8.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR