N-Methyldopamine hydrochloride 95%
Supplier: Thermo Scientific Chemicals
J60306-03
J60306-06
CAAAJ60306-03EA
700.48
CAD
CAAAJ60306-03
CAAAJ60306-06
N-Methyldopamine hydrochloride 95%
N-Methyldopamine hydrochloride
Dopamine agonist
Powder
Applications: Dopamine agonist
Formula:
C₉H₁₃NO₂·HCl Storage Temperature: Ambient |
MDL Number:
MFCD00035074 CAS Number: 62-32-8 EINECS: 200-528-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...