Nitrazine yellow

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 3-((2,4-dinitrophenyl)diazenyl)-4-hydroxynaphthalene-2,7-disulfonate, Sodium 3-((2,4-dinitrophenyl)diazenyl)-4-hydroxynaphthalene-2,7-disulphonate

J60281-03 J60281-06 J60281-14
CAAAJ60281-03EA 49.39 CAD
CAAAJ60281-03 CAAAJ60281-06 CAAAJ60281-14
Nitrazine yellow
Nitrazine yellow

pH indicator


Powder


Applications: pH indicator

Formula: C₁₆H₈N₄Na₂O₁₁S₂
MW: 542.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003938
CAS Number: 5423-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR