1-(4-Chlorophenyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

J60278-03 J60278-06
CAAAJ60278-03EA 83.65 CAD
CAAAJ60278-03 CAAAJ60278-06
1-(4-Chlorophenyl)piperazine 97%
1-(4-Chlorophenyl)piperazine

A 5-HT-1 serotonin receptor agonist


Purity: 97% CAS No.: 38212-33-8 Molecular Formula: C10H13ClN2 Formula Weight: 196.70 Physical Form: Solid Appearance: White Melting Point: 76-80° Boiling Point: 113° MDL No.: MFCD00044823


A 5-HT-1 serotonin receptor agonist

Formula: C₁₀H₁₃ClN₂
MW: 196.68
Boiling Pt: 113 °C
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00044823
CAS Number: 38212-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR