AMPD, 0.2 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60916-AE
J60916-AK
CAAAJ60276-AEEA
157.94
CAD
CAAAJ60276-AE
CAAAJ60276-AK
CAAAJ60916-AE
CAAAJ60916-AK
CAAAJ61377-AE
CAAAJ61377-AK
AMPD, 0.2 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
2-Amino-2-methyl-1,3-propanediol buffer, colorless and odorless.
CAS No.: 115-69-5
MDL No.: MFCD00004678
Caution: Storage Temperature: 2 to 8 °C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...