FSB buffer, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60262-AK
J60262-EQE
CAAAJ60262-AKEA
463.81
CAD
CAAAJ60262-AK
CAAAJ60262-EQE
FSB buffer, Thermo Scientific
Buffers
Cell Culture Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
For frozen storage of competent cells
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...