L(+)-Ornithine hydrochloride cell culture reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Orn-OH Hydrochloride, H-Orn-OH·HCl
J60241-18
J60241-22
J60241-30
CAAAJ60241-18EA
141.47
CAD
CAAAJ60241-18
CAAAJ60241-22
CAAAJ60241-30
L(+)-Ornithine hydrochloride cell culture reagent
L(+)-Ornithine hydrochloride
White crystalline powder
CAS No.: 3184-13-2
Molecular Formula: C5H12N2O2.HCl
Formula Weight: 168.62
Physical Form: Crystalline powder
Appearance: White
Melting Point: ca 238° dec.
Beilstein Reference: 3625847
Merck Reference: 14,6874
Optical Rotation: +23° (c=5 in 5N HCl)
MDL No.: MFCD00064562
Formula:
C₅H₁₂N₂O₂·HCl Melting Pt: ca 238 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064562 CAS Number: 3184-13-2 EINECS: 221-678-6 Merck Index: 14,6874 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...