TE buffer, pH 7.4, RNAse free, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60234-AK
J60234-EQE
CAAAJ60234-AKEA
174.84
CAD
CAAAJ60234-AK
CAAAJ60234-EQE
TE buffer, pH 7.4, RNAse free, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
Appearance: Clear
MDL No.: MFCD00236359
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...