TRIS, 1.0 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60202-K2
J60381-K2
J60452-K2
J60636-K2
J61036-K2
J61579-K2
J61798-K2
J62084-K2
J62085-K2
J62186-K2
J62577-K2
J62657-K2
J62726-K2
J63831-K2
CAAAJ60202-K2EA
150.07
CAD
CAAAJ60202-K2
CAAAJ60381-K2
CAAAJ60452-K2
CAAAJ60636-K2
CAAAJ61036-K2
CAAAJ61066-K2
CAAAJ61579-K2
CAAAJ61798-K2
CAAAJ62084-K2
CAAAJ62085-K2
CAAAJ62186-K2
CAAAJ62577-K2
CAAAJ62657-K2
CAAAJ62726-K2
CAAAJ63831-K2
TRIS, 1.0 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 77-86-1
Odor: Odorless
MDL No.: MFCD00004679
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...