Sodium orthovanadate 100 mM in aqueous solution

Supplier: Thermo Scientific Chemicals

J60191-AD J60191-AE
CAAAJ60191-ADEA 150.07 CAD
CAAAJ60191-AD CAAAJ60191-AE
Sodium orthovanadate 100 mM in aqueous solution
Sodium orthovanadate

Liquid


CAS No.: 13721-39-6 Molecular Formula: Na3O4V Formula Weight: 183.94 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Colorless Odor: Odorless MDL No.: MFCD00003511

Formula: Na₃O₄V
MW: 183.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003511
CAS Number: 13721-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR