Loratadine ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)piperidine-1-carboxylate

J60190-03 J60190-06 J60190-ME
CAAAJ60190-03EA 377.95 CAD
CAAAJ60190-03 CAAAJ60190-06 CAAAJ60190-ME
Loratadine ≥98%
Loratadine

A nonsedating-type histamine H1-receptor antagonist


Powder


Applications: A nonsedating-type histamine H1-receptor antagonist

Formula: C₂₂H₂₃ClN₂O₂
MW: 382.88 g/mol
Melting Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00672869
CAS Number: 79794-75-5
Merck Index: 14,5578

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR