MOPS-EDTA-sodium acetate buffer (MESA), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60184-AP
J60184-K2
CAAAJ60184-APEA
151.33
CAD
CAAAJ60184-AP
CAAAJ60184-K2
MOPS-EDTA-sodium acetate buffer (MESA), Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00131625
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...