Docetaxel 99%

Supplier: Thermo Scientific Chemicals
Warning

J60174-03 J60174-MC J60174-ME
CAAAJ60174-03EA 2267.51 CAD
CAAAJ60174-03 CAAAJ60174-MC CAAAJ60174-ME
Docetaxel 99%
Docetaxel

A second-generation cytotoxic antimicrotubule agent


Powder


Applications: A second-generation cytotoxic antimicrotubule agent

Formula: C₄₃H₅₃NO₁₄
Melting Pt: 186…192 °C
Storage Temperature: Freezer
MDL Number: MFCD00800737
CAS Number: 114977-28-5
Merck Index: 14,3397

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR