N-Decanoyl-N-methyl-D-glucamine (MEGA-10)

Supplier: Thermo Scientific Chemicals

J60173-06 J60173-03
CAAAJ60173-06EA 993.09 CAD
CAAAJ60173-06 CAAAJ60173-03
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
N-Decanoyl-N-methyl-D-glucamine (MEGA-10)

Non-ionic detergent for solubilizing membrane proteins.


Powder


Non-ionic detergent for solubilizing membrane proteins

Formula: C₁₇H₃₅NO₆
Melting Pt: 91…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036801
CAS Number: 85261-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR