Glycine, 0.2 M buffer soln., pH 3.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60154-AK
J60154-AP
CAAAJ60154-AKEA
150.07
CAD
CAAAJ60154-AK
CAAAJ60154-AP
Glycine, 0.2 M buffer soln., pH 3.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 56-40-6
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00008131
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...