NP-40 lysis buffer with glycerol (2X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60143-AK
J60143-EQE
CAAAJ60143-AKEA
242.85
CAD
CAAAJ60143-AK
CAAAJ60143-EQE
NP-40 lysis buffer with glycerol (2X), Thermo Scientific
Buffers
Extraction Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Viscous liquid
Appearance: Colorless
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...