Everolimus
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60139-LB0
J60139-MA
J60139-MB
CAAAJ60139-LB0EA
437.93
CAD
CAAAJ60139-LB0
CAAAJ60139-MA
CAAAJ60139-MB
Everolimus
Everolimus
A rapamycin analog
CAS No.: 159351-69-6
Molecular Formula: C53H83NO14
Formula Weight: 958.22
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
Solubility: Soluble in DMSO or ethanol
Merck Reference: 14,3907
MDL No.: MFCD07785165
A rapamycin analog
Formula:
C₅₃H₈₃NO₁₄ Storage Temperature: Freezer |
MDL Number:
MFCD00929329 CAS Number: 159351-69-6 Merck Index: 14,3907 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...