2-Iminothiolane hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-IT HCl, 2-Iminothiolane hydrochloride, 2-Thiolanimine hydrochloride, 2-IT
J60131-03
J60131-MD
CAAAJ60131-03EA
617.84
CAD
CAAAJ60131-03
CAAAJ60131-MD
2-Iminothiolane hydrochloride
2-Iminothiolane hydrochloride (Traut's Reagent)
Protein modification and cross-linking reagent, cleavable by thiols
Powder
Applications: Protein modification and cross-linking reagent, cleavable by thiols
Formula:
C₄H₈ClNS Melting Pt: 190…195 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00039013 CAS Number: 4781-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...