Alcian blue 8GX
Supplier: Thermo Scientific Chemicals
|
Warning
|
J60122-09
J60122-14
J60122-22
CAAAJ60122-09EA
239.28
CAD
CAAAJ60122-09
CAAAJ60122-14
CAAAJ60122-22
Alcian blue 8GX
Alcian blue 8 GX
Powder
CAS No.: 33864-99-2
Molecular Formula: C56H68Cl4CuN16S4
Formula Weight: 1298.88
Physical Form: Powder
Appearance: Dark blue
Melting Point: 148°
MDL No.: MFCD00010720
Formula:
C₅₆H₆₈Cl₄CuN₁₆S₄ Melting Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010720 CAS Number: 33864-99-2 EINECS: 251-705-7 Merck Index: 14,218 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...