Spironolactone

Supplier: Thermo Scientific Chemicals
Danger

J60119-03 J60119-06 J60119-09
CAAAJ60119-03EA 4578.21 CAD
CAAAJ60119-03 CAAAJ60119-06 CAAAJ60119-09
Spironolactone
Spironolactone

Antihypertensive potassium sparing diuretic; aldosterone receptor antagonist


Powder


Antihypertensive potassium sparing diuretic; aldosterone receptor antagonist

Formula: C₂₄H₃₂O₄S
MW: 416.58 g/mol
Boiling Pt: 597
Melting Pt: 196
Storage Temperature: Ambient
MDL Number: MFCD00082250
CAS Number: 52-01-7
EINECS: 200-133-6
Merck Index: 14,8760

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR