Tricine, 0.5 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60107-AE
J60107-AK
J62234-AE
J62234-AK
J62672-AE
J62672-AK
J62983-AE
J62983-AK
J63108-AE
J63108-AK
CAAAJ60107-AEEA
4628.37
CAD
CAAAJ60107-AE
CAAAJ60107-AK
CAAAJ62234-AE
CAAAJ62234-AK
CAAAJ62672-AE
CAAAJ62672-AK
CAAAJ62983-AE
CAAAJ62983-AK
CAAAJ63108-AE
CAAAJ63108-AK
CAAAJ63880-AE
CAAAJ63880-AK
Tricine, 0.5 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
N-(Trishydroxymethylmethyl)glycine buffer solutions, colorless and odorless.
CAS No.: 5704-04-1
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00004277
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...