Acetate, 1 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60104-AK J60104-AP J60340-AK J60340-AP J60964-AK J60964-AP J61033-AK J61033-AP J63876-AK J63876-AP
CAAAJ60104-AKEA 4347.04 CAD
CAAAJ60104-AK CAAAJ60104-AP CAAAJ60340-AK CAAAJ60340-AP CAAAJ60964-AK CAAAJ60964-AP CAAAJ61033-AK CAAAJ61033-AP CAAAJ63876-AK CAAAJ63876-AP
Acetate, 1 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 127-09-3
MDL No.: MFCD00012459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR