MOPS, 1.0 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60097-AK J61797-AK J61797-AP J61821-AK J61821-AP J61843-AK J61843-AP J61958-AK J61958-AP J62261-AK J62261-AP J62840-AK J62840-AP
CAAAJ60097-AKEA 1919.07 CAD
CAAAJ60097-AK CAAAJ60097-AP CAAAJ61797-AK CAAAJ61797-AP CAAAJ61821-AK CAAAJ61821-AP CAAAJ61843-AK CAAAJ61843-AP CAAAJ61958-AK CAAAJ61958-AP CAAAJ62261-AK CAAAJ62261-AP CAAAJ62682-AK CAAAJ62682-AP CAAAJ62840-AK CAAAJ62840-AP
MOPS, 1.0 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 1132-61-2
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00006183

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR