Chlortetracycline hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J60095-14 J60095-22
CAAAJ60095-14EA 202.24 CAD
CAAAJ60095-14 CAAAJ60095-22
Chlortetracycline hydrochloride
Chlortetracycline hydrochloride

Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria


Powder


Bacteriostatic antibiotic active against Gram-positive and Gram-negative bacteria

Formula: C₂₂H₂₃ClN₂O₈·HCl
Melting Pt: 210…215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD26142699
CAS Number: 64-72-2
EINECS: 200-591-7
Merck Index: 14,2192

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR