Bicarbonate, 1 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60092-AP J60116-AK
CAAAJ60092-AKEA 5085.64 CAD
CAAAJ60092-AK CAAAJ60092-AP CAAAJ60116-AK CAAAJ60116-AP CAAAJ63491-AK CAAAJ63491-AP
Bicarbonate, 1 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 144-55-8
MDL No.: MFCD00003528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR