Hydroxylapatite, fast flow
Supplier: Thermo Scientific Chemicals
Synonyms:
Calcium Phosphate, Tribasic
J60076-09
J60076-18
CAAAJ60076-09EA
146.78
CAD
CAAAJ60076-09
CAAAJ60076-18
Hydroxylapatite, fast flow
Hydroxylapatite
Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids.
Powder
Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids
Formula:
Ca₅HO₁₃P₃ MW: 502.32 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00010904 CAS Number: 1306-06-5 EINECS: 215-145-7 Merck Index: 14,3467 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...